missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%, Alfa Aesar™

$208.49 - $632.03

Chemical Identifiers

CAS 136705-64-1
Molecular Formula C22H36P2
Molecular Weight (g/mol) 362.478
MDL Number MFCD00142335
InChI Key GVVCHDNSTMEUCS-UAFMIMERSA-N
Synonym --1,2-bis 2r,5r-2,5-diethylphospholano benzene, --duphos, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, 7n2eb15paq, ethyl-duphos, r,r, r,r-et-duphos, r,r-et-duphos mi, r,r-ethyl-duphos, unii-7n2eb15paq
PubChem CID 2734549
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60470MD
View Documents
Thermo Fisher Scientific
H60470MD
250mg Each for $208.49
 
AAH6047003
View Documents
Thermo Fisher Scientific
H6047003
1g Each for $632.03
 
Specifications

Chemical Identifiers

136705-64-1
362.478
GVVCHDNSTMEUCS-UAFMIMERSA-N
2734549
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
C22H36P2
MFCD00142335
--1,2-bis 2r,5r-2,5-diethylphospholano benzene, --duphos, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, 7n2eb15paq, ethyl-duphos, r,r, r,r-et-duphos, r,r-et-duphos mi, r,r-ethyl-duphos, unii-7n2eb15paq
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane

Specifications

1.01
−265° (c=1 in Hexane)
Air Sensitive
136705-64-1
MFCD00142335
--1,2-bis 2r,5r-2,5-diethylphospholano benzene, --duphos, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, 7n2eb15paq, ethyl-duphos, r,r, r,r-et-duphos, r,r-et-duphos mi, r,r-ethyl-duphos, unii-7n2eb15paq
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
362.478
362.47
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene
>110°C (230°F)
1.6
250mg
C22H36P2
4814174
GVVCHDNSTMEUCS-UAFMIMERSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
2734549
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen