missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6047003

 View more versions of this product

Catalog No. AAH6047003



Specifications

Specifications

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene
−265° (c=1 in Hexane)
136705-64-1
MFCD00142335
ethyl-duphos, r,r, r,r-ethyl-duphos, unii-7n2eb15paq, --1,2-bis 2r,5r-2,5-diethylphospholano benzene, r,r-et-duphos, 7n2eb15paq, --1,2-bis 2r,5r-2,5-diethylphospholano benzene, --duphos, r,r-et-duphos mi, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
362.478
362.47
>110°C (230°F)
1g
C22H36P2
4814174
GVVCHDNSTMEUCS-UAFMIMERSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
2734549
≥97%

Chemical Identifiers

136705-64-1
362.478
GVVCHDNSTMEUCS-UAFMIMERSA-N
2734549
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
C22H36P2
MFCD00142335
ethyl-duphos, r,r, r,r-ethyl-duphos, unii-7n2eb15paq, --1,2-bis 2r,5r-2,5-diethylphospholano benzene, r,r-et-duphos, 7n2eb15paq, --1,2-bis 2r,5r-2,5-diethylphospholano benzene, --duphos, r,r-et-duphos mi, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents