Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis[2-(trifluoromethyl)phenyl]ethane, 97+%

$36.77 - $133.35

Chemical Identifiers

Chemical Name or Material 1,2-Bis[2-(trifluoromethyl)phenyl]ethane
CAS 287172-66-1
Molecular Formula C16H12F6
Formula Weight 318.26
MDL Number MFCD01631639
Synonym 2,2'-Bis(trifluoromethyl)dibenzyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2453303

  SDS
alfa aesar™
B2453303
1g Each for $36.77

AAB2453306

  SDS
alfa aesar™
B2453306
5g Each for $133.35
Description & Specifications

Specifications

Formula Weight 318.26
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥97%
Chemical Name or Material 1,2-Bis[2-(trifluoromethyl)phenyl]ethane
CAS 287172-66-1
Molecular Formula C16H12F6
MDL Number MFCD01631639
Quantity 1g
Melting Point 74° to 76°C
Synonym 2,2'-Bis(trifluoromethyl)dibenzyl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.