Please login to your online account to display your discounted pricing

1,2-Bis(2-chloroethoxy)ethane, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  349620050

 View more versions of this product

Catalog No. AC349620050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 187.07
Assay ≥96.0%
Quantity 5mL
Color Clear colorless to pale yellow
TSCA TSCA
RTECS Number KH4900000
EINECS Number 203-952-7
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Synonym Triethylene glycol dichloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Molecular Formula C6H12Cl2O2
MDL Number MFCD00000976
Name Note 97%
Linear Formula (-CH2OCH2CH2Cl)2
Chemical Name or Material 1, 2-Bis(2-chloroethoxy)ethane
CAS 112-26-5
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 1, 2-Bis(2-chloroethoxy)ethane
Name Note 97%
CAS 112-26-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12Cl2O2
Linear Formula (-CH2OCH2CH2Cl)2
Formula Weight 187.07
MDL Number MFCD00000976
Synonym Triethylene glycol dichloride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

SDS & Safety Documents