Please login to your online account to display your discounted pricing

1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic Acid Tetrasodium Salt 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  401650050

 View more versions of this product

Catalog No. AC401650050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 564.36
Quantity 5g
Color Beige
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Synonym BAPTA tetrasodium salt
Chemical Name or Material 1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid tetrasodium salt
Name Note 95%
Assay Percent Range 95%
Molecular Formula C22H20N2Na4O10
MDL Number MFCD00067691
CAS 126824-24-6
Health Hazard 1 LIGHT SENSITIVE
Health Hazard 2 MOISTURE SENSITIVE
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid tetrasodium salt
Name Note 95%
CAS 126824-24-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C22H20N2Na4O10
Formula Weight 564.36
MDL Number MFCD00067691
Synonym BAPTA tetrasodium salt
Accessories

LIGHT SENSITIVE

MOISTURE SENSITIVE

SDS & Safety Documents