missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane, TCI America™

$39.72 - $175.70

Chemical Identifiers

CAS 84852-53-9
Molecular Formula C14H4Br10
Molecular Weight (g/mol) 971.226
InChI Key BZQKBFHEWDPQHD-UHFFFAOYSA-N
Synonym 2,2′,3,3′,4,4′,5,5′,6,6′-Decabromobibenzyl
PubChem CID 10985889
IUPAC Name 1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
SMILES C(CC1=C(C(=C(C(=C1Br)Br)Br)Br)Br)C2=C(C(=C(C(=C2Br)Br)Br)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B279525G View Documents TCI America
B279525G
25g Each for $39.72
 
B2795500G View Documents TCI America
B2795500G
500g Each for $175.70
 
Specifications

Chemical Identifiers

84852-53-9
971.226
2,2′,3,3′,4,4′,5,5′,6,6′-Decabromobibenzyl
1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
C14H4Br10
BZQKBFHEWDPQHD-UHFFFAOYSA-N
10985889
C(CC1=C(C(=C(C(=C1Br)Br)Br)Br)Br)C2=C(C(=C(C(=C2Br)Br)Br)Br)Br

Specifications

Undesignated
25g
C14H4Br10
BZQKBFHEWDPQHD-UHFFFAOYSA-N
1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
10985889
Crystal-Powder at 20°C
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane
352°C
84852-53-9
2,2′C,3,3′C,4,4′C,5,5′C,6,6′C-Decabromobibenzyl
C(CC1=C(C(=C(C(=C1Br)Br)Br)Br)Br)C2=C(C(=C(C(=C2Br)Br)Br)Br)Br
971.226
971.23
≥96.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : DA0358200

TSCA : Yes