Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(1-naphthyl)ethane, 97%

$149.31 - $638.80

Chemical Identifiers

Chemical Name or Material 1,2-Bis(1-naphthyl)ethane
CAS 15374-45-5
Molecular Formula C22H18
Formula Weight 282.39
MDL Number MFCD00037736
Synonym 1,2-Di(1-naphthyl)ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2060903

 
alfa aesar™
B2060903
1g Each for $149.31

AAB2060906

 
alfa aesar™
B2060906
5g Each for $638.80
Description & Specifications

Specifications

Formula Weight 282.39
Chemical Name or Material 1,2-Bis(1-naphthyl)ethane
Assay 97%
Molecular Formula C22H18
MDL Number MFCD00037736
CAS 15374-45-5
Synonym 1,2-Di(1-naphthyl)ethane
Melting Point 161° to 162°C
Quantity 1g