missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Benzisoxazole, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2340003

 View more versions of this product

Catalog No. AAB2340003


Description

Description

1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. 4.1 mg/mL in water at 25°C

Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
Specifications

Specifications

1,2-Benzisoxazole
1.169
86°C (186°F)
1.562
1 g
Light sensitive
Soluble in water. 4.1mg/mL in water at 25°C
C1=CC=C2C(=C1)C=NO2
119.123
CHEBI:51554
97%
271-95-4
90°C to 92°C (15 mmHg)
C7H5NO
MFCD00005852
2154
1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine
KTZQTRPPVKQPFO-UHFFFAOYSA-N
1,2-benzoxazole
71073
119.12

Chemical Identifiers

271-95-4
119.123
KTZQTRPPVKQPFO-UHFFFAOYSA-N
71073
1,2-benzoxazole
C7H5NO
MFCD00005852
1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine
CHEBI:51554
C1=CC=C2C(=C1)C=NO2
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 205-983-1

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only