missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Benzisoxazol-3-amine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451710250
Specifications
1, 2-Benzisoxazol-3-amine | |
110°C to 112°C | |
36216-80-5 | |
C7H6N2O | |
NLMVYUBGWZWUGB-UHFFFAOYSA-N | |
1,2-benzoxazol-3-amine | |
2779749 | |
97% |
Glass bottle | |
25g | |
96% min. (HPLC) | |
1,2-benzisoxazol-3-amine, 1,2-benzoisoxazol-3-amine, 3-amino-1,2-benzisoxazole, 3-aminobenzisoxazole, amino-benzoisoxazole, aminobenzisoxazole, benzo d isoxazol-3-amine, benzo d isoxazol-3-ylamine, benzo d isoxazole-3-ylamine, zlchem 93 | |
C1=CC=C2C(=C1)C(=NO2)N | |
134.14 | |
134.14 |
Chemical Identifiers
36216-80-5 | |
134.14 | |
1,2-benzisoxazol-3-amine, 1,2-benzoisoxazol-3-amine, 3-amino-1,2-benzisoxazole, 3-aminobenzisoxazole, amino-benzoisoxazole, aminobenzisoxazole, benzo d isoxazol-3-amine, benzo d isoxazol-3-ylamine, benzo d isoxazole-3-ylamine, zlchem 93 | |
1,2-benzoxazol-3-amine |
C7H6N2O | |
NLMVYUBGWZWUGB-UHFFFAOYSA-N | |
2779749 | |
C1=CC=C2C(=C1)C(=NO2)N |