missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Benzenedithiol, TCI America™

$161.95 - $559.17

Chemical Identifiers

CAS 17534-15-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.234
MDL Number MFCD00004835
InChI Key JRNVQLOKVMWBFR-UHFFFAOYSA-N
Synonym 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
PubChem CID 69370
IUPAC Name benzene-1,2-dithiol
SMILES C1=CC=C(C(=C1)S)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13221G View Documents TCI America
B13221G
1g Each for $161.95
 
B13225G View Documents TCI America
B13225G
5g Each for $559.17
 
Specifications

Chemical Identifiers

17534-15-5
142.234
JRNVQLOKVMWBFR-UHFFFAOYSA-N
69370
C1=CC=C(C(=C1)S)S
C6H6S2
MFCD00004835
1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
benzene-1,2-dithiol

Specifications

239°C
1g
C6H6S2
1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
C1=CC=C(C(=C1)S)S
142.234
142.23
≥95.0% (GC)
White-Yellow
17534-15-5
MFCD00004835
JRNVQLOKVMWBFR-UHFFFAOYSA-N
benzene-1,2-dithiol
69370
Crystal-Lump at 20°C
1,2-Benzenedithiol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator