missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Benzenedisulfonyl Dichloride, TCI America™

$157.36 - $1,825.73

Chemical Identifiers

CAS 6461-76-3
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.114
MDL Number MFCD00059836
InChI Key YBGQXNZTVFEKEN-UHFFFAOYSA-N
Synonym benzene-1,2-disulfonyl dichloride, 1,2-benzenedisulfonyl dichloride, o-benzenedisulfonyl chloride, 1,2-benzenedisulfonyldichloride, 1,2-benzenedisulfonyl chloride, acmc-209nnc, benzenedisulfonyl chloride, benzenedisulfonyl dichloride, benzene-1,2-disulfochloride, 1,2-benzenedisulphonyl chloride
PubChem CID 246432
IUPAC Name benzene-1,2-disulfonyl chloride
SMILES C1=CC=C(C(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11281G View Documents TCI America
B11281G
1g Each for $157.36
 
B11285G View Documents TCI America
B11285G
5g Each for $525.56
 
B112825G View Documents TCI America
B112825G
25g Each for $1,825.73
 
Specifications

Chemical Identifiers

6461-76-3
275.114
YBGQXNZTVFEKEN-UHFFFAOYSA-N
246432
C1=CC=C(C(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl
C6H4Cl2O4S2
MFCD00059836
benzene-1,2-disulfonyl dichloride, 1,2-benzenedisulfonyl dichloride, o-benzenedisulfonyl chloride, 1,2-benzenedisulfonyldichloride, 1,2-benzenedisulfonyl chloride, acmc-209nnc, benzenedisulfonyl chloride, benzenedisulfonyl dichloride, benzene-1,2-disulfochloride, 1,2-benzenedisulphonyl chloride
benzene-1,2-disulfonyl chloride

Specifications

White-Yellow
1g
C6H4Cl2O4S2
3261
YBGQXNZTVFEKEN-UHFFFAOYSA-N
benzene-1,2-disulfonyl chloride
246432
Crystal-Powder at 20°C
1,2-Benzenedisulfonyl Dichloride
144°C
6461-76-3
MFCD00059836
benzene-1,2-disulfonyl dichloride, 1,2-benzenedisulfonyl dichloride, o-benzenedisulfonyl chloride, 1,2-benzenedisulfonyldichloride, 1,2-benzenedisulfonyl chloride, acmc-209nnc, benzenedisulfonyl chloride, benzenedisulfonyl dichloride, benzene-1,2-disulfochloride, 1,2-benzenedisulphonyl chloride
C1=CC=C(C(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl
275.114
275.11
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No