missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Aminophenyl)pyrrole, 98+%, ACROS Organics™

Chemical Identifiers

CAS 6025-60-1
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C10 H10 N2
Formula Weight 158.2
MDL Number MFCD00005344
Chemical Name or Material 1-(2-Aminophenyl)pyrrole, 98+%
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC188500050 SDS Thermo Fisher Scientific
188500050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6025-60-1
100.0
C10 H10 N2
MFCD00005344
≥98.0%
98.0
98+%
158.2
1-(2-Aminophenyl)pyrrole, 98+%

Specifications

Crystalline Powder and Needles
5g
94° to 98°C
100.0
Glass bottle
6025-60-1
Authentic
≥98.0%
158.2
Light beige to brown
98.0
MFCD00005344
C10 H10 N2
98+%
1-(2-Aminophenyl)pyrrole, 98+%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 227-884-2

TSCA : TSCA