Learn More
1-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2464506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(2-Aminoethyl)pyrrolidine | |
0.904 | |
47°C (116°F) | |
C6H14N2 | |
MFCD00003182 | |
UN2734 | |
Air sensitive; Hygroscopic | |
WRXNJTBODVGDRY-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylethanamine | |
1344 | |
99% |
7154-73-6 | |
161°C | |
Amine-like | |
1.4687 | |
5 g | |
102978 | |
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine | |
C1CCN(C1)CCN | |
114.192 | |
114.19 |
Chemical Identifiers
7154-73-6 | |
114.192 | |
WRXNJTBODVGDRY-UHFFFAOYSA-N | |
1344 | |
C1CCN(C1)CCN |
C6H14N2 | |
MFCD00003182 | |
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine | |
2-pyrrolidin-1-ylethanamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 230-509-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only