Learn More
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 400760025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(2-Aminoethyl)piperazine | |
140-31-8 | |
100.0 | |
218.0°C to 222.0°C | |
Authentic | |
Glass bottle | |
1.4990 to 1.5010 | |
2.5kg | |
n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl | |
IMUDHTPIFIBORV-UHFFFAOYSA-N | |
2-piperazin-1-ylethanamine | |
8795 | |
129.2 |
99% | |
98.5 | |
0.9800g/mL | |
88°C | |
98.5% min. (GC) | |
C6H15N3 | |
MFCD00005971 | |
0.98 | |
Solubility in water: soluble. | |
NCCN1CCNCC1 | |
129.21 | |
14 mPa.s (20°C) | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
GHS Signal Word: Danger
EINECSNumber : 205-411-0
RTECSNumber : TK8050000
TSCA : TSCA
RUO â Research Use Only