missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Aminoethyl)piperazine, 98%

Manufacturer:  Alfa Aesar™ A1015430

 View more versions of this product

Catalog No. AAA1015430



Description

Description

Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
Specifications

Specifications

1- (2-Aminoethyl)piperazine
222°C
250g
C6H15N3
UN2815
n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
C1CN(CCN1)CCN
129.207
129.21
93°C (199°F)
-19°C
140-31-8
MFCD00005971
104363
IMUDHTPIFIBORV-UHFFFAOYSA-N
2-piperazin-1-ylethanamine
8795
98%

Chemical Identifiers

140-31-8
129.207
IMUDHTPIFIBORV-UHFFFAOYSA-N
8795
C1CN(CCN1)CCN
C6H15N3
MFCD00005971
n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
2-piperazin-1-ylethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H317-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: N-AMINOETHYLPIPERAZINE

RTECSNumber : TK8050000

TSCA : TSCA

SDS Documents