Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Aminoethyl)-4-methylhomopiperazine, 98%

Chemical Identifiers

Chemical Name or Material 1-(2-Aminoethyl)-4-methylhomopiperazine
CAS 39270-45-6
Molecular Formula C8H19N3
Formula Weight 157.26
MDL Number MFCD08061018
UN Number UN2735
Synonym 2-(4-Methyl-1,4-diazepan-1-yl)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5083703

  SDS
alfa aesar™
H5083703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50837MD

  SDS
alfa aesar™
H50837MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.26
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD08061018
Synonym 2-(4-Methyl-1,4-diazepan-1-yl)ethylamine
Molecular Formula C8H19N3
Assay 98%
CAS 39270-45-6
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 1 Danger
UN Number UN2735
Chemical Name or Material 1-(2-Aminoethyl)-4-methylhomopiperazine
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.