missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 213030500

 View more versions of this product

Catalog No. AC213030500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

261.11
100.0
2HCl·H2O
MFCD00066965
98%
50g
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate
49575-10-2
98%
97.5

Chemical Identifiers

49575-10-2
100.0
2HCl·H2O
MFCD00066965
97.5
98%
261.11
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 256-385-2

TSCA : TSCA