missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98%, ACROS Organics™
Chemical Identifiers
CAS | 49575-10-2 |
---|---|
Molecular Formula | 2HCl·H2O |
Molecular Weight (g/mol) | 261.11 |
MDL Number | MFCD00066965 |
InChI Key | AZYQXHDJVHNBRE-UHFFFAOYSA-N |
Synonym | dihydrogen 2-2-methyl-5-nitroimidazol-1-yl ethanamine hydrate dichloride |
PubChem CID | 12222573 |
IUPAC Name | 2-(2-methyl-5-nitroimidazol-1-yl)ethanamine;dihydrochloride |
SMILES | CC1=NC=C(N1CCN)[N+](=O)[O-].Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC213030050
|
Thermo Fisher Scientific
213030050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
49575-10-2 | |
261.11 | |
AZYQXHDJVHNBRE-UHFFFAOYSA-N | |
12222573 | |
CC1=NC=C(N1CCN)[N+](=O)[O-].Cl.Cl |
2HCl·H2O | |
MFCD00066965 | |
dihydrogen 2-2-methyl-5-nitroimidazol-1-yl ethanamine hydrate dichloride | |
2-(2-methyl-5-nitroimidazol-1-yl)ethanamine;dihydrochloride |
Specifications
Glass bottle | |
5g | |
97.5 | |
98% | |
MFCD00066965 | |
Solubility in water: 600g/L. Other solubilities: methanol:26g/L ethanol:1.8g/L 2-propanol:0.48g/l,dichloromethane: 0.11g/L,acetone: 0.11g/L,n,n-dimethylformamide: 7g/L | |
CC1=NC=C(N1CCN)[N+](=O)[O-].Cl.Cl | |
261.11 | |
261.11 | |
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98% |
195°C | |
49575-10-2 | |
100.0 | |
2HCl·H2O | |
dihydrogen 2-2-methyl-5-nitroimidazol-1-yl ethanamine hydrate dichloride | |
AZYQXHDJVHNBRE-UHFFFAOYSA-N | |
2-(2-methyl-5-nitroimidazol-1-yl)ethanamine;dihydrochloride | |
12222573 | |
98% |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
GHS Signal Word: Warning
EINECSNumber : 256-385-2
TSCA : TSCA