missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, 98%, Alfa Aesar™

$54.74 - $862.04

Chemical Identifiers

CAS 130198-05-9
Molecular Formula C15H24ClNO2
Molecular Weight (g/mol) 285.812
MDL Number MFCD06658143
InChI Key NTKXIDDUCSFBBF-UHFFFAOYSA-N
Synonym 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride, 2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride, 1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride, n,n-didesmethylvenlafaxine hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride, 1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
PubChem CID 9795856
IUPAC Name 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
SMILES COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6486206 View Documents Alfa Aesar
H6486206
5g Each for $54.74
 
AAH6486214 View Documents Alfa Aesar
H6486214
25g Each for $215.89
 
AAH6486222 View Documents Alfa Aesar
H6486222
100g Each for $862.04
 
Specifications

Chemical Identifiers

130198-05-9
285.812
NTKXIDDUCSFBBF-UHFFFAOYSA-N
9795856
COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl
C15H24ClNO2
MFCD06658143
1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride, 2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride, 1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride, n,n-didesmethylvenlafaxine hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride, 1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride

Specifications

White
5g
C15H24ClNO2
4215444
NTKXIDDUCSFBBF-UHFFFAOYSA-N
1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
9795856
Crystalline Powder
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
174°C to 178°C (literature)
130198-05-9
MFCD06658143
1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride, 2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride, 1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride, n,n-didesmethylvenlafaxine hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride, 1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl
285.812
285.81
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning