missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 866926-59-2
Molecular Formula C24H33N2·Cl
Molecular Weight (g/mol) 385
InChI Key RHFFYNDQCMNENO-UHFFFAOYSA-M
Synonym 1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride
PubChem CID 11617932
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
SMILES CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC435101000 SDS View Documents Thermo Fisher Scientific
AC435101000
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435105000 View Documents Thermo Fisher Scientific
AC435105000
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

866926-59-2
385
1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride
1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
C24H33N2·Cl
RHFFYNDQCMNENO-UHFFFAOYSA-M
11617932
CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]

Specifications

Authentic
Violet to White
100mg
96.0
96% min. (HPLC)
1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride
CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]
385
385
97%
Glass Bottle
318°C to 319°C
866926-59-2
100.0
C24H33N2·Cl
RHFFYNDQCMNENO-UHFFFAOYSA-M
1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
11617932
Powder
1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning