Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,6-Dichlorophenyl)ethanol, 97%

$48.82 - $48.82

Chemical Identifiers

Chemical Name or Material 1-(2,6-Dichlorophenyl)ethanol
CAS 53066-19-6
Molecular Formula C8H8Cl2O
Formula Weight 191.06
MDL Number MFCD00016853
Beilstein 3243202
Synonym 2,6-Dichloro-alpha-methylbenzyl alcohol; 2,6-Dichlorophenyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1235003

  SDS
alfa aesar™
L1235003
1g Each for $48.82
Description & Specifications

Specifications

Formula Weight 191.06
Chemical Name or Material 1-(2,6-Dichlorophenyl)ethanol
Molecular Formula C8H8Cl2O
Assay 97%
Synonym 2,6-Dichloro-alpha-methylbenzyl alcohol; 2,6-Dichlorophenyl methyl carbinol
MDL Number MFCD00016853
CAS 53066-19-6
Beilstein 3243202
Boiling Point 117° to 118°C (4mmHg)
Quantity 1g