Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,5-Trichloro-3-iodobenzene, 97%

$43.55 - $329.41

Chemical Identifiers

Chemical Name or Material 1,2,5-Trichloro-3-iodobenzene
CAS 216393-66-7
Molecular Formula C6H2Cl3I
Formula Weight 307.35
MDL Number MFCD01318175
Beilstein 3128014
Synonym 1-Iodo-2,3,5-trichlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1765703

 
alfa aesar™
L1765703
1g Each for $43.55

AAL1765706

 
alfa aesar™
L1765706
5g Each for $105.89

AAL1765714

 
alfa aesar™
L1765714
25g Each for $329.41
Description & Specifications

Specifications

Formula Weight 307.35
Assay 97%
Molecular Formula C6H2Cl3I
CAS 216393-66-7
Beilstein 3128014
Melting Point 50° to 54°C
Chemical Name or Material 1,2,5-Trichloro-3-iodobenzene
MDL Number MFCD01318175
Synonym 1-Iodo-2,3,5-trichlorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.