missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,5-Trichloro-3-iodobenzene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1765714

 View more versions of this product

Catalog No. AAL1765714



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2,5-Trichloro-3-iodobenzene
50°C to 54°C
MFCD01318175
3128014
1-iodo-2,3,5-trichlorobenzene, 1,2,5-trichloro-3-iodo-benzene, acmc-1clah, benzene,1,2,5-trichloro-3-iodo, benzene, 1,2,5-trichloro-3-iodo
C1=C(C=C(C(=C1Cl)Cl)I)Cl
307.336
307.35
216393-66-7
C6H2Cl3I
25 g
Light sensitive
YLLMVDOSVGVBJC-UHFFFAOYSA-N
1,2,5-trichloro-3-iodobenzene
20269860
97%

Chemical Identifiers

216393-66-7
307.336
YLLMVDOSVGVBJC-UHFFFAOYSA-N
1,2,5-trichloro-3-iodobenzene
C6H2Cl3I
MFCD01318175
20269860
C1=C(C=C(C(=C1Cl)Cl)I)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only