Please login to your online account to display your discounted pricing

1-(2,5-Dimethylphenyl)piperazine 99%, ACROS Organics™

$59.32 - $59.32

Chemical Identifiers

Chemical Name or Material 1-(2, 5-Dimethylphenyl)piperazine
Name Note 99%
CAS 1013-25-8
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5% total base, ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C12 H18 N2
Formula Weight 190.29
MDL Number MFCD00038378
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC136950050

  SDS
thermo fisher scientific
136950050
Glass bottle 5g Each for $59.32
Description & Specifications

Specifications

Assay 98.5 to 101.5% total base, ≥98.5% (GC)
Physical Form Crystalline Solid
Formula Weight 190.29
Quantity 5g
Color White
EINECS Number 213-795-6
TSCA TSCA
Melting Point 42° to 46°C
CAS Max % 100.0
CAS Min % 98.5
Chemical Name or Material 1-(2, 5-Dimethylphenyl)piperazine
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C12 H18 N2
MDL Number MFCD00038378
CAS 1013-25-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.