Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,5-Dimethylphenyl)piperazine, 96%

Chemical Identifiers

Chemical Name or Material 1-(2,5-Dimethylphenyl)piperazine
CAS 1013-25-8
Molecular Formula C12H18N2
Formula Weight 190.28
MDL Number MFCD00038378
UN Number UN3259
Beilstein 611229
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0574406

 
alfa aesar™
L0574406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0574414

 
alfa aesar™
L0574414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.28
UN Number UN3259
Chemical Name or Material 1-(2,5-Dimethylphenyl)piperazine
Beilstein 611229
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Molecular Formula C12H18N2
Melting Point 42° to 47°C
Boiling Point 154°C (10mmHg)
MDL Number MFCD00038378
CAS 1013-25-8
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.