Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,5-Dimethoxybenzyl)piperazine hydrochloride, 97%

$60.66 - $197.03

Chemical Identifiers

Chemical Name or Material 1-(2,5-Dimethoxybenzyl)piperazine hydrochloride
CAS 374897-99-1
Molecular Formula C13H20N2O2·HCl
Formula Weight 272.78
MDL Number MFCD03093088
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1911009

  SDS
alfa aesar™
L1911009
10g Each for $60.66

AAL1911018

  SDS
alfa aesar™
L1911018
50g Each for $197.03
Description & Specifications

Specifications

Formula Weight 272.78
Chemical Name or Material 1-(2,5-Dimethoxybenzyl)piperazine hydrochloride
Assay 97%
Molecular Formula C13H20N2O2·HCl
MDL Number MFCD03093088
CAS 374897-99-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 141° to 143°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.