missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC270965000
Specifications
1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose | |
Glass bottle | |
+ 37.5 | |
73°C to 77°C | |
2595-05-3 | |
100.0 | |
C12H20O6 | |
1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-bis-o-1-methylethylidene, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 5,6-di-o-isopropylidene-alpha-d-allofuranose | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C | |
260.28 | |
260.28 | |
98% |
Authentic | |
+37.50 (20.00°C c=1,chloroform) | |
White | |
500mg | |
97.5 | |
97.5% min. (GC) | |
MFCD00135634 | |
KEJGAYKWRDILTF-VVULQXIFSA-N | |
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol | |
7157054 | |
Powder |
Chemical Identifiers
2595-05-3 | |
260.28 | |
KEJGAYKWRDILTF-VVULQXIFSA-N | |
7157054 | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C |
C12H20O6 | |
MFCD00135634 | |
1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-bis-o-1-methylethylidene, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 5,6-di-o-isopropylidene-alpha-d-allofuranose | |
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol |