Please login to your online account to display your discounted pricing

1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
CAS 2595-05-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C12H20O6
Formula Weight 260.28
MDL Number MFCD00135634
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC270965000

  SDS
thermo fisher scientific
AC270965000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC270960100

  SDS
thermo fisher scientific
AC270960100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC270960025

  SDS
thermo fisher scientific
270960025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Powder
Formula Weight 260.28
Quantity 500mg
Color White to off-white
Specific Rotation Condition +37.50 (20.00°C c=1,chloroform)
Melting Point 72° to 77°C
CAS Max % 100.0
Assay Percent Range 98%
CAS 2595-05-3
Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
Molecular Formula C12H20O6
MDL Number MFCD00135634
Packaging Glass bottle
Specific Rotation +33.0° to +39.0° (20°C, 589nm) (c=1, CHCl3)
CAS Min % 97.5
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.