missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 270960025

 View more versions of this product

Catalog No. AC270960025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
Glass bottle
+ 37.5
73°C to 77°C
2595-05-3
100.0
C12H20O6
1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-bis-o-1-methylethylidene, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 5,6-di-o-isopropylidene-alpha-d-allofuranose
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C
260.28
260.28
98%
Authentic
+37.50 (20.00°C c=1,chloroform)
White
2.5g
97.5
97.5% min. (GC)
MFCD00135634
KEJGAYKWRDILTF-VVULQXIFSA-N
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
7157054
Powder

Chemical Identifiers

2595-05-3
260.28
KEJGAYKWRDILTF-VVULQXIFSA-N
7157054
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C
C12H20O6
MFCD00135634
1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-bis-o-1-methylethylidene, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 5,6-di-o-isopropylidene-alpha-d-allofuranose
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SDS Documents