Learn More
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140090100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,4-Trimethylbenzene | |
97.5 | |
-44.0°C | |
0.8800g/mL | |
48°C | |
98% | |
C9H12 | |
C6H3(CH3)3 | |
10 L | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and ether | |
CC1=CC(=C(C=C1)C)C | |
120.195 | |
CHEBI:34039 | |
98% |
95-63-6 | |
100.0 | |
Colorless | |
168.0°C | |
Authentic | |
Plastic drum | |
1.5030 to 1.5050 | |
MFCD00008527 | |
pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene | |
GWHJZXXIDMPWGX-UHFFFAOYSA-N | |
1,2,4-trimethylbenzene | |
7247 | |
120.19 | |
Liquid |
Chemical Identifiers
95-63-6 | |
GWHJZXXIDMPWGX-UHFFFAOYSA-N | |
7247 | |
1,2,4-trimethylbenzene |
MFCD00008527 | |
pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene | |
CHEBI:34039 | |
CC1=CC(=C(C=C1)C)C |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only