Learn More
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012411B4
Description
1,2,4-Trimethylbenzene is applied as a liquid scintillator. It is also used as a sterilizing agent and in the manufacture of dyes, perfumes, and resins. It is used as a safe solvent for the production of H2O2 (hydrogen peroxide). Pseudocumene is used mainly as raw material for the production of trimellitic anhydride, pyromellitic dianhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,4-Trimethylbenzene | |
-44°C | |
169°C to 171°C | |
Aromatic | |
1.504 | |
16 kg | |
1903005 | |
pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene | |
GWHJZXXIDMPWGX-UHFFFAOYSA-N | |
1,2,4-trimethylbenzene | |
7247 | |
120.2 | |
Liquid |
95-63-6 | |
0.882 g/mL | |
48°C (118°F) | |
C9H12 | |
MFCD00008527 | |
UN3295 | |
14,7915 | |
Soluble in alcohol,benzene and ether; Slightly soluble in water | |
CC1=CC(=C(C=C1)C)C | |
120.195 | |
CHEBI:34039 | |
98% |
Chemical Identifiers
95-63-6 | |
120.195 | |
GWHJZXXIDMPWGX-UHFFFAOYSA-N | |
7247 | |
1,2,4-trimethylbenzene |
C9H12 | |
MFCD00008527 | |
pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene | |
CHEBI:34039 | |
CC1=CC(=C(C=C1)C)C |
Safety and Handling
GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 202-436-9
RTECSNumber : DC3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only