Please login to your online account to display your discounted pricing

1,2,4-Trifluoro-5-nitrobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2, 4-Trifluoro-5-nitrobenzene
CAS 2105-61-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6 H2 F3 N O2
Linear Formula F3C6H2NO2
Formula Weight 177.08
MDL Number MFCD00007051
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303730010

  SDS
thermo fisher scientific
303730010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 177.08
Quantity 1g
Color Clear brown
Density 1.5400g/mL
CAS Max % 100.0
Chemical Name or Material 1, 2, 4-Trifluoro-5-nitrobenzene
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Molecular Formula C6 H2 F3 N O2
CAS 2105-61-5
MDL Number MFCD00007051
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula F3C6H2NO2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.