Please login to your online account to display your discounted pricing

1,2,4-Trifluoro-5-nitrobenzene, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  303730010

 View more versions of this product

Catalog No. AC303730010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 177.08
Physical Form Liquid
Quantity 1g
Color Clear brown
Density 1.5400g/mL
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 1, 2, 4-Trifluoro-5-nitrobenzene
Molecular Formula C6 H2 F3 N O2
Assay Percent Range 99%
CAS 2105-61-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00007051
CAS Min % 98.5
Linear Formula F3C6H2NO2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

Specifications

Chemical Name or Material 1, 2, 4-Trifluoro-5-nitrobenzene
CAS 2105-61-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6 H2 F3 N O2
Linear Formula F3C6H2NO2
Formula Weight 177.08
MDL Number MFCD00007051
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

SDS & Safety Documents