Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4-Trifluoro-5-nitrobenzene, 99%

Chemical Identifiers

Chemical Name or Material 1,2,4-Trifluoro-5-nitrobenzene
CAS 2105-61-5
Molecular Formula C6H2F3NO2
Formula Weight 177.08
MDL Number MFCD00007051
UN Number UN2810
Beilstein 2504370
Synonym 1-Nitro-2,4,5-trifluorobenzene; 2,4,5-Trifluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1657906

 
alfa aesar™
A1657906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1657903

 
alfa aesar™
A1657903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1657914

 
alfa aesar™
A1657914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.08
Melting Point −11°C
Boiling Point 195°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Chemical Name or Material 1,2,4-Trifluoro-5-nitrobenzene
Assay 99%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Flash Point 89°C (192°F)
UN Number UN2810
Molecular Formula C6H2F3NO2
MDL Number MFCD00007051
CAS 2105-61-5
Beilstein 2504370
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Nitro-2,4,5-trifluorobenzene; 2,4,5-Trifluoronitrobenzene
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.