Learn More
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.01%
$62.64 - $569.37
Chemical Identifiers
CAS | 120-82-1 |
---|---|
Molecular Formula | C6H3Cl3 |
Molecular Weight (g/mol) | 181.44 |
MDL Number | MFCD00000547 |
InChI Key | PBKONEOXTCPAFI-UHFFFAOYSA-N |
Synonym | benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene |
PubChem CID | 13 |
ChEBI | CHEBI:28222 |
IUPAC Name | 1,2,4-trichlorobenzene |
SMILES | C1=CC(=C(C=C1Cl)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA19390AK
|
Thermo Scientific Chemicals
019390AK |
250 mL |
Each for $62.64
|
|
|||||
AA19390K2
|
Thermo Scientific Chemicals
019390K2 |
1 L |
Each for $180.58
|
|
|||||
AA19390K7
|
Thermo Scientific Chemicals
019390K7 |
4 L |
Each for $569.37
|
|
|||||
Description
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-82-1 | |
181.44 | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
13 | |
1,2,4-trichlorobenzene |
C6H3Cl3 | |
MFCD00000547 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
CHEBI:28222 | |
C1=CC(=C(C=C1Cl)Cl)Cl |
Specifications
17°C | |
Colorless | |
213°C | |
110°C (230°F) | |
Aromatic | |
1.572 | |
C6H3Cl3 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
1,2,4-trichlorobenzene | |
13 | |
181.45 | |
1, 2, 4-Trichlorobenzene |
120-82-1 | |
1.454 | |
250 mL | |
571°C (1060°F) | |
Liquid | |
99% min. | |
MFCD00000547 | |
Immiscible with water. | |
C1=CC(=C(C=C1Cl)Cl)Cl | |
181.44 | |
CHEBI:28222 | |
Spectrophotometric | |
1, 2, 4-Trichlorobenzene |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 204-428-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only