Learn More
1,2,4-Trichlorobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A10130CLL
Description
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
1,2,4-Trichlorobenzene | |
120-82-1 | |
213°C | |
110°C (230°F) | |
1.572 | |
MFCD00000547 | |
14,9631 | |
Miscible with ether,benzene,CS2;Immiscible with water. | |
C1=CC(=C(C=C1Cl)Cl)Cl | |
181.44 | |
CHEBI:28222 | |
99% |
17°C | |
1.454g/mL | |
4 x 2.5 kg | |
Liquid | |
C6H3Cl3 | |
UN2321 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
1,2,4-trichlorobenzene | |
13 | |
181.45 |
Chemical Identifiers
120-82-1 | |
181.44 | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
13 | |
1,2,4-trichlorobenzene |
C6H3Cl3 | |
MFCD00000547 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
CHEBI:28222 | |
C1=CC(=C(C=C1Cl)Cl)Cl |
Safety and Handling
GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TRICHLOROBENZENES, LIQUID
EINECSNumber : 204-428-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only