Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4-Trichloro-5-iodobenzene, 98%

$78.35 - $280.15

Chemical Identifiers

Chemical Name or Material 1,2,4-Trichloro-5-iodobenzene
CAS 7145-82-6
Molecular Formula C6H2Cl3I
Formula Weight 307.35
MDL Number MFCD00051450
Beilstein 1940881
Synonym 1-Iodo-2,4,5-trichlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0601506

  SDS
alfa aesar™
L0601506
5g Each for $78.35

AAL0601514

  SDS
alfa aesar™
L0601514
25g Each for $280.15
Description & Specifications

Specifications

Formula Weight 307.35
Chemical Name or Material 1,2,4-Trichloro-5-iodobenzene
Assay 98%
Molecular Formula C6H2Cl3I
MDL Number MFCD00051450
Beilstein 1940881
Synonym 1-Iodo-2,4,5-trichlorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 294°C (110mmHg)
Melting Point 102° to 105°C
Quantity 5g
CAS 7145-82-6

Applications
1,2,4-Trichloro-5-iodobenzene can be used to produce 3-(2,4,5-trichloro-phenyl)-acrylic acid tert-butyl ester at the temperature of 80°C.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.