Learn More
1,2,4-Triazole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1159736
Description
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and bases.
Specifications
1,2,4-Triazole | |
118°C to 122°C | |
158°C (316°F) | |
MFCD00005228 | |
104767 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
1H-1,2,4-triazole | |
9257 | |
69.07 |
288-88-0 | |
260°C | |
C2H3N3 | |
500 g | |
14,9605 | |
Soluble in water. | |
C1=NC=NN1 | |
69.067 | |
CHEBI:35550 | |
99% |
Chemical Identifiers
288-88-0 | |
69.067 | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
9257 | |
1H-1,2,4-triazole |
C2H3N3 | |
MFCD00005228 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
CHEBI:35550 | |
C1=NC=NN1 |
Safety and Handling
GHS H Statement
H361-H302-H319
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes serious eye irritation.
P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
H302-H319-H361d
EINECSNumber : 206-022-9
RTECSNumber : XZ3806000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only