missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,4-Triazole-3-carboxamide, TCI America™

Manufacturer:  TCI America T240225G

 View more versions of this product

Catalog No. T240225G



Specifications

Specifications

1,2,4-Triazole-3-carboxamide
315°C
3641-08-5
MFCD03990481
C1=NNC(=N1)C(=O)N
112.09
112.09
≥97.0% (HPLC,N)
White-Yellow
25g
C3H4N4O
ZEWJFUNFEABPGL-UHFFFAOYSA-N
1H-1,2,4-triazole-5-carboxamide
65125
Crystal-Powder at 20°C

Chemical Identifiers

3641-08-5
112.09
ZEWJFUNFEABPGL-UHFFFAOYSA-N
1H-1,2,4-triazole-5-carboxamide
C3H4N4O
MFCD03990481
65125
C1=NNC(=N1)C(=O)N
Safety and Handling

Safety and Handling

TSCA : No

Documents