Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4-Triaminobenzene dihydrochloride, 96%

$138.54 - $467.95

Chemical Identifiers

Chemical Name or Material 1,2,4-Triaminobenzene dihydrochloride
CAS 615-47-4
Molecular Formula C6H9N3·2HCl
Formula Weight 196.08
MDL Number MFCD00016619
Synonym 1,2,4-Benzenetriamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1775806

 
alfa aesar™
A1775806
5g Each for $138.54

AAA1775814

 
alfa aesar™
A1775814
25g Each for $467.95
Description & Specifications

Specifications

Formula Weight 196.08
Chemical Name or Material 1,2,4-Triaminobenzene dihydrochloride
TSCA TSCA
Assay 96%
Molecular Formula C6H9N3·2HCl
MDL Number MFCD00016619
CAS 615-47-4
Synonym 1,2,4-Benzenetriamine dihydrochloride
Melting Point ≈290°C (dec.)
Health Hazard 1 Danger
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DC1953000

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.