Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4-Triacetoxybenzene, 97%

$38.49 - $114.99

Chemical Identifiers

Chemical Name or Material 1,2,4-Triacetoxybenzene
CAS 613-03-6
Molecular Formula C12H12O6
Formula Weight 252.22
MDL Number MFCD00008700
Beilstein 2138876
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2420814

 
alfa aesar™
B2420814
25g Each for $38.49

AAB2420822

 
alfa aesar™
B2420822
100g Each for $114.99
Description & Specifications

Specifications

Formula Weight 252.22
Chemical Name or Material 1,2,4-Triacetoxybenzene
TSCA TSCA
Assay 97%
Molecular Formula C12H12O6
MDL Number MFCD00008700
CAS 613-03-6
Beilstein 2138876
Melting Point 98° to 100°C
Quantity 25g
RTECS Number DC4800000