Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[2-(4-Morpholinyl)ethyl]piperazine, 97%

$108.34 - $470.11

Chemical Identifiers

Chemical Name or Material 1-[2-(4-Morpholinyl)ethyl]piperazine
CAS 4892-89-1
Molecular Formula C10H21N3O
Formula Weight 199.3
MDL Number MFCD00191217
Synonym 4-[2-(1-Piperazinyl)ethyl]morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068803

  SDS
alfa aesar™
H5068803
1g Each for $108.34

AAH5068806

  SDS
alfa aesar™
H5068806
5g Each for $470.11
Description & Specifications

Specifications

Formula Weight 199.3
Chemical Name or Material 1-[2-(4-Morpholinyl)ethyl]piperazine
Assay 97%
Molecular Formula C10H21N3O
MDL Number MFCD00191217
CAS 4892-89-1
Synonym 4-[2-(1-Piperazinyl)ethyl]morpholine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 106° to 107°C (0.2mmHg)
Melting Point 36° to 38°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.