Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4-Dichlorobenzyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2,4-Dichlorobenzyl)piperazine
CAS 51619-56-8
Formula Weight 245.15
MDL Number MFCD03407487
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5544003

 
alfa aesar™
H5544003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5544006

 
alfa aesar™
H5544006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5544014

 
alfa aesar™
H5544014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.15
Chemical Name or Material 1-(2,4-Dichlorobenzyl)piperazine
Refractive Index 1.5710
Physical Form Liquid
Assay 97%
MDL Number MFCD03407487
CAS 51619-56-8
Density 1.255g/mL
Color Pale yellow
Quantity 1g