missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4-Dichlorobenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5544014

 View more versions of this product

Catalog No. AAH5544014



Specifications

Specifications

1-(2,4-Dichlorobenzyl)piperazine
1.5710
25g
C11H14Cl2N2
1-2,4-dichloro-benzyl-piperazine
C1CN(CCN1)CC2=C(C=C(C=C2)Cl)Cl
245.147
245.15
97%
1.255g/mL
Yellow
51619-56-8
MFCD03407487
VYKXBWXIOSLDQR-UHFFFAOYSA-N
1-[(2,4-dichlorophenyl)methyl]piperazine
411179
Liquid

Chemical Identifiers

51619-56-8
245.147
VYKXBWXIOSLDQR-UHFFFAOYSA-N
411179
C1CN(CCN1)CC2=C(C=C(C=C2)Cl)Cl
C11H14Cl2N2
MFCD03407487
1-2,4-dichloro-benzyl-piperazine
1-[(2,4-dichlorophenyl)methyl]piperazine
Documents