missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4-Dichlorobenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5544006

 View more versions of this product

Catalog No. AAH5544006



Specifications

Specifications

1-(2,4-Dichlorobenzyl)piperazine
>110°C (230°F)
Yellow
51619-56-8
MFCD03407487
VYKXBWXIOSLDQR-UHFFFAOYSA-N
1-[(2,4-dichlorophenyl)methyl]piperazine
411179
Liquid
1.255 g/mL
1.571
5g
C11H14Cl2N2
1-2,4-dichloro-benzyl-piperazine
C1CN(CCN1)CC2=C(C=C(C=C2)Cl)Cl
245.147
245.15
97%

Chemical Identifiers

51619-56-8
245.147
VYKXBWXIOSLDQR-UHFFFAOYSA-N
411179
C1CN(CCN1)CC2=C(C=C(C=C2)Cl)Cl
C11H14Cl2N2
MFCD03407487
1-2,4-dichloro-benzyl-piperazine
1-[(2,4-dichlorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents