Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2,4-Butanetriol, 96%

$20.51 - $20.51

Chemical Identifiers

Chemical Name or Material 1,2,4-Butanetriol
CAS 3068-00-6
Molecular Formula C4H10O3
Formula Weight 106.12g/mol
MDL Number MFCD00002929
Beilstein 1733456
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0513806

 
alfa aesar™
L05138-06
5g Each for $20.51
Description & Specifications

Specifications

Quantity 5g
Melting Point −20°C
Boiling Point 190° to 191°C (18mmHg)
Quantity 5g
Flash Point 188°C (370°F)
Chemical Name or Material 1,2,4-Butanetriol
CAS 3068-00-6
Assay 96%
Molecular Formula C4H10O3
Formula Weight 106.12g/mol
MDL Number MFCD00002929
Beilstein 1733456
RTECS Number EK7176000
TSCA TSCA