Please login to your online account to display your discounted pricing

1,2,4-Benzenetricarboxylic Anhydride 97%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  105370250

 View more versions of this product

Catalog No. AC105370250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 192.13
Physical Form Flakes
Quantity 25g
Color White to off-white
TSCA TSCA
EINECS Number 209-008-0
Melting Point 164° to 169°C
RTECS Number DC2050000
Beilstein 18, 468
Merck Index 15, 9872
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1, 2, 4-Benzenetricarboxylic anhydride
Infrared Spectrum Authentic
Name Note 97%
Packaging Plastic bottle
Assay Percent Range 97%
Molecular Formula C9H4O5
MDL Number MFCD00005925
CAS 552-30-7
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Trimellitic anhydride
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

Specifications

Chemical Name or Material 1, 2, 4-Benzenetricarboxylic anhydride
Name Note 97%
CAS 552-30-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H4O5
Formula Weight 192.13
MDL Number MFCD00005925
Synonym Trimellitic anhydride
Accessories

MOISTURE SENSITIVE

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

SDS & Safety Documents