Please login to your online account to display your discounted pricing

1,2,4-Benzenetricarboxylic Anhydride 97%, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  105370010

 View more versions of this product

Catalog No. AC105370010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Flakes
Formula Weight 192.13
Quantity 1kg
Color White to off-white
Beilstein 18, 468
Merck Index 15, 9872
EINECS Number 209-008-0
TSCA TSCA
RTECS Number DC2050000
Melting Point 164° to 169°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1, 2, 4-Benzenetricarboxylic anhydride
Assay Percent Range 97%
Molecular Formula C9H4O5
MDL Number MFCD00005925
CAS 552-30-7
Health Hazard 1 MOISTURE SENSITIVE
Infrared Spectrum Authentic
Name Note 97%
Synonym Trimellitic anhydride
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

Specifications

Chemical Name or Material 1, 2, 4-Benzenetricarboxylic anhydride
Name Note 97%
CAS 552-30-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H4O5
Formula Weight 192.13
MDL Number MFCD00005925
Synonym Trimellitic anhydride
Accessories

MOISTURE SENSITIVE

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

SDS & Safety Documents