missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4,6-Trimethylbenzyl)piperazine, 97%

$71.53 - $969.14

Chemical Identifiers

CAS 41717-26-4
Molecular Formula C14H22N2
Molecular Weight (g/mol) 218.344
MDL Number MFCD03094587
InChI Key FOERNUXLFALRDN-UHFFFAOYSA-N
Synonym 1-2 4 6-trimethylbenzyl piperazine
PubChem CID 2737385
IUPAC Name 1-[(2,4,6-trimethylphenyl)methyl]piperazine
SMILES CC1=CC(=C(C(=C1)C)CN2CCNCC2)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5573403 View Documents Alfa Aesar™
H5573403
1g Each for $71.53
 
AAH5573406 View Documents Alfa Aesar™
H5573406
5g Each for $234.65
 
AAH5573414 View Documents Alfa Aesar™
H5573414
25g Each for $969.14
 
Specifications

Chemical Identifiers

41717-26-4
218.344
FOERNUXLFALRDN-UHFFFAOYSA-N
2737385
CC1=CC(=C(C(=C1)C)CN2CCNCC2)C
C14H22N2
MFCD03094587
1-2 4 6-trimethylbenzyl piperazine
1-[(2,4,6-trimethylphenyl)methyl]piperazine

Specifications

White to Yellow
1g
C14H22N2
1-2 4 6-trimethylbenzyl piperazine
CC1=CC(=C(C(=C1)C)CN2CCNCC2)C
218.344
218.34
97%
88°C to 98°C
41717-26-4
MFCD03094587
FOERNUXLFALRDN-UHFFFAOYSA-N
1-[(2,4,6-trimethylphenyl)methyl]piperazine
2737385
Powder
1-(2,4,6-Trimethylbenzyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures