Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole, 98%

$30.45 - $298.34

Chemical Identifiers

Chemical Name or Material 1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole
CAS 54230-60-3
Molecular Formula C17H25N3O2S
Formula Weight 335.47
MDL Number MFCD00014089
Synonym 1-(2,4,6-Triisopropylbenzenesulfonyl)-1,2,4-triazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2503803

  SDS
alfa aesar™
B2503803
1g Each for $30.45

AAB2503806

  SDS
alfa aesar™
B2503806
5g Each for $79.72

AAB2503814

  SDS
alfa aesar™
B2503814
25g Each for $298.34
Description & Specifications

Specifications

Formula Weight 335.47
Assay 98%
Synonym 1-(2,4,6-Triisopropylbenzenesulfonyl)-1,2,4-triazole
Molecular Formula C17H25N3O2S
Quantity 1g
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
MDL Number MFCD00014089
Melting Point 110° to 112°C
Chemical Name or Material 1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 54230-60-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C